missing translation for 'onlineSavingsMsg'
Learn More

Dibromoisocyanuric Acid 97.0+%, TCI America™

Catalog No. p-7127894
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37535G 5 g
D375325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37535G Supplier TCI America Supplier No. D37535G

Chemical Identifiers

CAS 15114-43-9
Molecular Formula C3HBr2N3O3
Molecular Weight (g/mol) 286.867
MDL Number MFCD00463941
InChI Key HHBCEKAWSILOOP-UHFFFAOYSA-N
Synonym 1,3-Dibromo-1,3,5-triazine-2,4,6-trione, DBI
PubChem CID 192912
IUPAC Name 1,3-dibromo-1,3,5-triazinane-2,4,6-trione
SMILES C1(=O)NC(=O)N(C(=O)N1Br)Br

Specifications

Melting Point 309°C
Quantity 5 g
UN Number 3085
Formula Weight 286.87
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dibromoisocyanuric Acid
TSCA No
Recommended Storage Refrigerator