accessibility menu, dialog, popup

UserName

BrNONHONBrO

CAS RN 15114-43-9

IUPAC Name: 1,3-dibromo-1,3,5-triazinane-2,4,6-trione
Synonyms: 1,3-dibromo-1,3,5-triazinane-2,4,6-trione
Molecular Weight (g/mol): 286.87
Molecular Formula: C3HBr2N3O3
InChi Key: HHBCEKAWSILOOP-UHFFFAOYSA-N
SMILES: BrN1C(=O)NC(=O)N(Br)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results