missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibromochloromethane (stabilized with Ethanol) 95.0+%, TCI America™
$103.97 - $340.75
Chemical Identifiers
| CAS | 124-48-1 |
|---|---|
| Molecular Formula | CHBr2Cl |
| Molecular Weight (g/mol) | 208.277 |
| MDL Number | MFCD00000820 |
| InChI Key | GATVIKZLVQHOMN-UHFFFAOYSA-N |
| Synonym | chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform |
| PubChem CID | 31296 |
| ChEBI | CHEBI:34627 |
| IUPAC Name | dibromo(chloro)methane |
| SMILES | C(Cl)(Br)Br |
Chemical Identifiers
| 124-48-1 | |
| 208.277 | |
| GATVIKZLVQHOMN-UHFFFAOYSA-N | |
| 31296 | |
| dibromo(chloro)methane |
| CHBr2Cl | |
| MFCD00000820 | |
| chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform | |
| CHEBI:34627 | |
| C(Cl)(Br)Br |
Specifications
| 124-48-1 | |
| 122°C | |
| MFCD00000820 | |
| 3077 | |
| GATVIKZLVQHOMN-UHFFFAOYSA-N | |
| dibromo(chloro)methane | |
| 31296 | |
| 208.28 | |
| Liquid |
| Yellow | |
| CHBr2Cl | |
| 5 g | |
| chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform | |
| C(Cl)(Br)Br | |
| 208.277 | |
| CHEBI:34627 | |
| ≥95.0% (GC) | |
| Dibromochloromethane (stabilized with Ethanol) |
Safety and Handling
RTECSNumber : PA6360000
TSCA : Yes