missing translation for 'onlineSavingsMsg'
Learn More

Dibromochloromethane (stabilized with Ethanol) 95.0+%, TCI America™

Catalog No. C08545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08545G 5 g
C085425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C08545G Supplier TCI America Supplier No. C08545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 124-48-1
Molecular Formula CHBr2Cl
Molecular Weight (g/mol) 208.277
MDL Number MFCD00000820
InChI Key GATVIKZLVQHOMN-UHFFFAOYSA-N
Synonym chlorodibromomethane, methane, dibromochloro, dibromo chloro methane, monochlorodibromomethane, cdbm, methane, chlorodibromo, unii-3t4ajr1h24, ccris 938, dibromo-chloro-methane, chlorobromoform
PubChem CID 31296
ChEBI CHEBI:34627
IUPAC Name dibromo(chloro)methane
SMILES C(Cl)(Br)Br

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 g
UN Number 3077
Formula Weight 208.28
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dibromochloromethane (stabilized with Ethanol)
RTECSNumber PA6360000
TSCA Yes