Learn More
Dibromoacetonitrile, 94%
CAS: 3252-43-5 | C2HBr2N | 198.845 g/mol
$62.32 - $451.92
Chemical Identifiers
| CAS | 3252-43-5 |
|---|---|
| Molecular Formula | C2HBr2N |
| Molecular Weight (g/mol) | 198.845 |
| MDL Number | MFCD00001856 |
| InChI Key | NDSBDLSWTGLNQA-UHFFFAOYSA-N |
| Synonym | dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 |
| PubChem CID | 18617 |
| ChEBI | CHEBI:82445 |
| IUPAC Name | 2,2-dibromoacetonitrile |
| SMILES | C(#N)C(Br)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1699406
|
Thermo Scientific Chemicals
A1699406 |
5 g |
Each for $62.32
|
|
|||||
|
AAA1699422
|
Thermo Scientific Chemicals
A1699422 |
100 g |
Each for $451.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3252-43-5 | |
| 198.845 | |
| NDSBDLSWTGLNQA-UHFFFAOYSA-N | |
| 18617 | |
| 2,2-dibromoacetonitrile |
| C2HBr2N | |
| MFCD00001856 | |
| dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 | |
| CHEBI:82445 | |
| C(#N)C(Br)Br |
Specifications
| 3252-43-5 | |
| 67°C to 69°C (24 mmHg) | |
| Pungent, Makes Eyes Water | |
| 1.542 | |
| MFCD00001856 | |
| UN3286 | |
| Light sensitive | |
| NDSBDLSWTGLNQA-UHFFFAOYSA-N | |
| 2,2-dibromoacetonitrile | |
| 18617 | |
| 198.84 | |
| Dibromoacetonitrile |
| 2.296 | |
| 56°C (133°F) | |
| C2HBr2N | |
| Br2CHCN | |
| 5 g | |
| 1739037 | |
| dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940 | |
| C(#N)C(Br)Br | |
| 198.845 | |
| CHEBI:82445 | |
| 94% |
Safety and Handling
GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
H301-H312+H332-H315-H319-H335-H351
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 221-843-2
RTECSNumber : AL8450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only