missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetonitrile, 94%

Catalog No. AAA1699406
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1699406 5 g
AAA1699422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1699406 Supplier Thermo Scientific Chemicals Supplier No. A1699406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3252-43-5 | C2HBr2N | 198.845 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3252-43-5
Molecular Formula C2HBr2N
Molecular Weight (g/mol) 198.845
MDL Number MFCD00001856
InChI Key NDSBDLSWTGLNQA-UHFFFAOYSA-N
Synonym dibromoacetonitrile, acetonitrile, dibromo, unii-vgj91h57xu, ccris 2673, 2,2-dibromoethanenitrile, vgj91h57xu, acetonitrile, 2,2-dibromo, dsstox_cid_4940, dsstox_rid_77590, dsstox_gsid_24940
PubChem CID 18617
ChEBI CHEBI:82445
IUPAC Name 2,2-dibromoacetonitrile
SMILES C(#N)C(Br)Br

Specifications

Density 2.296
Boiling Point 67°C to 69°C (24 mmHg)
Flash Point 56°C (133°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.542
Linear Formula Br2CHCN
Quantity 5 g
UN Number UN3286
Beilstein 1739037
Sensitivity Light sensitive
Formula Weight 198.84
Percent Purity 94%
Chemical Name or Material Dibromoacetonitrile
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H335-H351
Hazard Statement GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 221-843-2
RTECSNumber AL8450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only