missing translation for 'onlineSavingsMsg'
Learn More

Dibromoacetic acid, MilliporeSigma™ Supelco™

Numéro de catalogue. 111020081
Modifier l'affichage
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111020081 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111020081 Fournisseur Merck Emd Millipore Code fournisseur 442551
Il en reste null

Dibromoacetic acid belongs to the category of haloacetic acids (HAAs) which are a class of disinfection by-products produced in water.

Environmental

Spécifications

Packaging Ampule
CAS 631-64-1
Density 2.382 g/mL (at 25°C)
Flash Point 113°C
Physical Form Neat
Linear Formula Br2 CHCOOH
MDL Number MFCD00004205
Melting Point 32°C to 38°C
Boiling Point 128°C to 130°C (16 mmHg)
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 217.84
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats