missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl Sulfoxide 99.0+%, TCI America™
Supplier: TCI America D0001250G
Specifications
| Dibenzyl Sulfoxide | |
| 133°C | |
| C14H14OS | |
| 250 g | |
| HTMQZWFSTJVJEQ-UHFFFAOYSA-N | |
| (phenylmethanesulfinylmethyl)benzene | |
| 12116 | |
| ≥99.0% (HPLC) |
| 621-08-9 | |
| White-Yellow | |
| MFCD00004782 | |
| dibenzyl sulfoxide, benzyl sulfoxide, dibenzylsulfoxide, tardiol d, sulfoxide, dibenzyl, dibenzyl sulphoxide, sulfinylbis methylene dibenzene, bis phenylmethyl sulfoxide, nsc 55, benzene, 1,1'-sulfinylbis methylene bis | |
| O=S(CC1=CC=CC=C1)CC1=CC=CC=C1 | |
| 230.33 | |
| 230.33 | |
| Crystalline Powder |
Chemical Identifiers
| 621-08-9 | |
| 230.33 | |
| HTMQZWFSTJVJEQ-UHFFFAOYSA-N | |
| 12116 | |
| O=S(CC1=CC=CC=C1)CC1=CC=CC=C1 |
| C14H14OS | |
| MFCD00004782 | |
| dibenzyl sulfoxide, benzyl sulfoxide, dibenzylsulfoxide, tardiol d, sulfoxide, dibenzyl, dibenzyl sulphoxide, sulfinylbis methylene dibenzene, bis phenylmethyl sulfoxide, nsc 55, benzene, 1,1'-sulfinylbis methylene bis | |
| (phenylmethanesulfinylmethyl)benzene |
Safety and Handling
RTECSNumber : DA9275000
TSCA : Yes