missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl succinate, 98%

Catalog No. p-7028118
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2525714 25 g
AAB2525722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2525714 Supplier Thermo Scientific Chemicals Supplier No. B2525714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-43-5 | C18H18O4 | 298.34 g/mol

Dibenzyl succinate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl succinate is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 103-43-5
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.34
MDL Number MFCD00022034
InChI Key ODBOBZHTGBGYCK-UHFFFAOYSA-N
Synonym dibenzyl succinate, butanedioic acid, bis phenylmethyl ester, spasmine, benzyl succinate, dibenzylsuccinate, succinic acid dibenzyl ester, unii-ck14ug55av, ck14ug55av, diphenylmethyl butane-1,4-dioate, succinic acid, dibenzyl ester
PubChem CID 7653
ChEBI CHEBI:3058
IUPAC Name dibenzyl butanedioate
SMILES O=C(CCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Melting Point 42°C to 47°C
Density 1.256
Boiling Point 245°C (15 mmHg)
Refractive Index 1.596
Quantity 25 g
Beilstein 1916056
Solubility Information Insoluble in water.
Formula Weight 298.34
Percent Purity 98%
Chemical Name or Material Dibenzyl succinate
EINECSNumber 203-110-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only