accessibility menu, dialog, popup

UserName

OOOO

CAS RN 103-43-5

IUPAC Name: 1,4-dibenzyl butanedioate
Synonyms: dibenzyl succinate
Molecular Weight (g/mol): 298.34
Molecular Formula: C18H18O4
InChi Key: ODBOBZHTGBGYCK-UHFFFAOYSA-N
SMILES: O=C(CCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)