Learn More
Dibenzyl phosphite, 90+%
CAS: 17176-77-1 | C14H14O3P | 261.24 g/mol
Supplier: Thermo Scientific Chemicals L0985814
Description
Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.
Solubility
Soluble in DMSO, Methanol. Reacts with water.
Notes
Store at 4°C. Moisture Sensitive. Incompatible with moisture and oxidizing agents. Store under dry inert gas. Store away from oxidizing agents.
Specifications
| Dibenzyl phosphite | |
| 1.187 | |
| >110°C (230°F) | |
| C14H14O3P | |
| MFCD00004774 | |
| 1982794 | |
| 14,3014 | |
| Soluble in DMSO,Methanol. Reacts with water. | |
| O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1 | |
| 261.24 | |
| 262.25 |
| 17176-77-1 | |
| 110°C to 120°C (0.1 mmHg) | |
| Odorless | |
| 1.554 | |
| 25 g | |
| Moisture sensitive | |
| dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade | |
| RQKYHDHLEMEVDR-UHFFFAOYSA-N | |
| oxo-bis(phenylmethoxy)phosphanium | |
| 6334615 | |
| ≥90% |
Chemical Identifiers
| 17176-77-1 | |
| 261.24 | |
| RQKYHDHLEMEVDR-UHFFFAOYSA-N | |
| 6334615 | |
| O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1 |
| C14H14O3P | |
| MFCD00004774 | |
| dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade | |
| oxo-bis(phenylmethoxy)phosphanium |
Safety and Handling
GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 241-226-1
TSCA : No
Recommended Storage : Store under Nitrogen
RUO – Research Use Only