missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphite, 90+%

Catalog No. p-7045218
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0985806 5 g
AAL0985814 25 g
AAL0985822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0985806 Supplier Thermo Scientific Chemicals Supplier No. L0985806
Only null left

CAS: 17176-77-1 | C14H14O3P | 261.24 g/mol

Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

Solubility
Soluble in DMSO, Methanol. Reacts with water.

Notes
Store at 4°C. Moisture Sensitive. Incompatible with moisture and oxidizing agents. Store under dry inert gas. Store away from oxidizing agents.

Chemical Identifiers

CAS 17176-77-1
Molecular Formula C14H14O3P
Molecular Weight (g/mol) 261.24
MDL Number MFCD00004774
InChI Key RQKYHDHLEMEVDR-UHFFFAOYSA-N
Synonym dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
PubChem CID 6334615
IUPAC Name oxo-bis(phenylmethoxy)phosphanium
SMILES O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Specifications

Density 1.187
Boiling Point 110°C to 120°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.554
Quantity 5 g
Beilstein 1982794
Sensitivity Moisture sensitive
Merck Index 14,3014
Solubility Information Soluble in DMSO,Methanol. Reacts with water.
Formula Weight 262.25
Percent Purity ≥90%
Chemical Name or Material Dibenzyl phosphite
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 241-226-1
TSCA No
Recommended Storage Store under Nitrogen

RUO – Research Use Only