missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Phosphate 98.0+%, TCI America™

Supplier:  TCI America P11205G

 View more versions of this product

Catalog No. P11205G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dibenzyl Phosphate
81°C
C14H15O4P
5 g
HDFFVHSMHLDSLO-UHFFFAOYSA-N
dibenzyl hydrogen phosphate
74189
≥98.0% (HPLC,T)
1623-08-1
White
MFCD00004775
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2
278.244
278.24
Crystalline Powder

Chemical Identifiers

1623-08-1
278.244
HDFFVHSMHLDSLO-UHFFFAOYSA-N
74189
C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2
C14H15O4P
MFCD00004775
dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
dibenzyl hydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions