accessibility menu, dialog, popup

UserName

CAS RN 1623-08-1

IUPAC Name: bis(benzyloxy)phosphinic acid
Synonyms: dibenzyloxyphosphinic acid
Molecular Weight (g/mol): 278.24
Molecular Formula: C14H15O4P
InChi Key: HDFFVHSMHLDSLO-UHFFFAOYSA-N
SMILES: OP(=O)(OCC1=CC=CC=C1)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)