missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphate, 98%

Catalog No. AAL0881303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0881303 1 g
AAL0881306 5 g
AAL0881314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0881303 Supplier Thermo Scientific Chemicals Supplier No. L0881303

CAS: 1623-08-1 | C14H15O4P | 278.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1623-08-1
Molecular Formula C14H15O4P
Molecular Weight (g/mol) 278.244
MDL Number MFCD00004775
InChI Key HDFFVHSMHLDSLO-UHFFFAOYSA-N
Synonym dibenzyl phosphate, phosphoric acid dibenzyl ester, dibenzylphosphoric acid, unii-y65r35lz0s, phosphoric acid, bis phenylmethyl ester, dibenzylphosphate, creatine phosphate disodium impurity 4, dibenzyloxyphosphinic acid, dibenzyl-phosphate, pubchem14883
PubChem CID 74189
IUPAC Name dibenzyl hydrogen phosphate
SMILES C1=CC=C(C=C1)COP(=O)(O)OCC2=CC=CC=C2

Specifications

Melting Point 78°C to 81°C
Quantity 1 g
Beilstein 2055755
Formula Weight 278.24
Percent Purity 98%
Chemical Name or Material Dibenzyl phosphate
EINECSNumber 216-602-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only