missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl N,N-diisopropylphosphoramidite, 90+%

Catalog No. p-4813454
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC399640050 5 g
AC399640250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC399640050 Supplier Thermo Scientific Chemicals Supplier No. 399640050
Only null left

CAS: 108549-23-1 | C20H28NO2P | 345.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108549-23-1
Molecular Formula C20H28NO2P
Molecular Weight (g/mol) 345.42
InChI Key ANPWLBTUUNFQIO-UHFFFAOYSA-N
Synonym dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
PubChem CID 196621
IUPAC Name N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.0280g/mL
Flash Point 69°C
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.028
Formula Weight 345.42
Percent Purity 90+%
Chemical Name or Material Dibenzyl N, N-diisopropylphosphoramidite
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only