missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl L-Tartrate 97.0+%, TCI America™

Catalog No. T167125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T167125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T167125G Supplier TCI America Supplier No. T167125G
Only null left

Chemical Identifiers

CAS 622-00-4
Molecular Formula C18H18O6
Molecular Weight (g/mol) 330.336
MDL Number MFCD00674554
InChI Key LCKIPSGLXMCAOF-HZPDHXFCSA-N
Synonym L-Tartaric Acid Dibenzyl Ester
PubChem CID 7020934
IUPAC Name dibenzyl (2R,3R)-2,3-dihydroxybutanedioate
SMILES C1=CC=C(C=C1)COC(=O)C(C(C(=O)OCC2=CC=CC=C2)O)O

Specifications

Melting Point 68°C
Color White
Quantity 25 g
Formula Weight 330.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl L-Tartrate