accessibility menu, dialog, popup

UserName

OOOOOHOH

CAS RN 622-00-4

IUPAC Name: 1,4-dibenzyl 2,3-dihydroxybutanedioate
Synonyms: 1,4-dibenzyl 2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 330.34
Molecular Formula: C18H18O6
InChi Key: LCKIPSGLXMCAOF-UHFFFAOYNA-N
SMILES: OC(C(O)C(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results