missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl L-Glutamate Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128497
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48395G 5 g
D483925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48395G Supplier TCI America Supplier No. D48395G
Only null left

Chemical Identifiers

CAS 4561-10-8
Molecular Formula C19H22ClNO4
Molecular Weight (g/mol) 363.84
MDL Number MFCD00035103
InChI Key GRGJVECUQLAEDM-LMOVPXPDSA-N
Synonym s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl
PubChem CID 199613
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N.Cl

Specifications

Melting Point 104°C
Color White
Quantity 5 g
Formula Weight 363.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzyl L-Glutamate Hydrochloride
RTECSNumber MA0885000
TSCA No