missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl disulfide, 98+%

Catalog No. p-7045750
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1185014 25 g
AAL1185022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1185014 Supplier Thermo Scientific Chemicals Supplier No. L1185014
Only null left

CAS: 150-60-7 | C14H14S2 | 246.386 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 150-60-7
Molecular Formula C14H14S2
Molecular Weight (g/mol) 246.386
MDL Number MFCD00004783
InChI Key GVPWHKZIJBODOX-UHFFFAOYSA-N
Synonym dibenzyl disulfide, benzyl disulfide, disulfide, bis phenylmethyl, benzyl bisulfide, 1,2-dibenzyldisulfane, dibenzyl disulphide, dibenzyldisulfid, bis phenylmethyl disulfide, di phenylmethyl disulfide, disulfide, dibenzyl
PubChem CID 9012
ChEBI CHEBI:72752
IUPAC Name (benzyldisulfanyl)methylbenzene
SMILES C1=CC=C(C=C1)CSSCC2=CC=CC=C2

Specifications

Melting Point 67°C to 72°C
Density 1.3
Boiling Point 210°C to 216°C (18 mmHg)
Flash Point 150°C (302°F)
Odor Stench
Quantity 25 g
Beilstein 1110443
Merck Index 14,3013
Formula Weight 246.4
Percent Purity ≥98%
Chemical Name or Material Dibenzyl disulfide
Show More Show Less
Hazard Category H317
Hazard Statement GHS H Statement
H317
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 205-764-0
RTECSNumber JO1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only