Learn More
Dibenzyl diselenide, 95%
CAS: 1482-82-2 | C14H14Se2 | 340.208 g/mol
Supplier: Thermo Scientific Chemicals B2127803
Description
Dibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDibenzyl diselenide is used as a source of the PhSe unit in organic synthesis. It is also used to introduce PhSe groups by reaction with a variety of nucleophilic, including enolates, enol silyl ethers, Grignard reagents, organolithium reagents, alkenes and amines.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Specifications
| Dibenzyl diselenide | |
| 91°C to 93°C | |
| MFCD00004767 | |
| UN3283 | |
| dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene | |
| HYAVEDMFTNAZQE-UHFFFAOYSA-N | |
| (benzyldiselanyl)methylbenzene | |
| 95955 | |
| 95% |
| 1482-82-2 | |
| C14H14Se2 | |
| 1 g | |
| 1877745 | |
| Soluble in water. | |
| C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2 | |
| 340.208 | |
| 340.18 |
Chemical Identifiers
| 1482-82-2 | |
| 340.208 | |
| HYAVEDMFTNAZQE-UHFFFAOYSA-N | |
| 95955 | |
| C1=CC=C(C=C1)C[Se][Se]CC2=CC=CC=C2 |
| C14H14Se2 | |
| MFCD00004767 | |
| dibenzyl diselenide, 1,2-dibenzyldiselane, benzyl diselenide, dibenzyldiselenide, dibenzyldiselane, diselenide, bis phenylmethyl, dibenzyldiselenium, bisbenzyldiselenide, diselenide,bis phenylmethyl, benzyldiselanyl methylbenzene | |
| (benzyldiselanyl)methylbenzene |
Safety and Handling
GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only