missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl diisopropylphosphoramidite, 90+%

Catalog No. p-7026569
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2262906 5 g
AAB2262914 25 g
AAB2262922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2262906 Supplier Thermo Scientific Chemicals Supplier No. B2262906
Only null left

CAS: 108549-23-1 | C20H28NO2P | 345.423 g/mol

Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl N,N-diisopropylphosphoramidite may be used for the preparation of phosphopeptides. It is used for the synthesis of a GDP (guanosine diphosphate) analog, SML-8-73-1. It is useful for nucleotide coupling.

Solubility
Soluble in chloroform.

Notes
Moisture sensitive. Store under inert gas. Store at 0°C. Incompatible with oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 108549-23-1
Molecular Formula C20H28NO2P
Molecular Weight (g/mol) 345.423
MDL Number MFCD00191988
InChI Key ANPWLBTUUNFQIO-UHFFFAOYSA-N
Synonym dibenzyl diisopropylphosphoramidite, dibenzyl n,n-diisopropylphosphoramidite, bis benzyloxy diisopropylamino phosphine, dibenzyldiisopropylphosphoramidite, dibenzyloxy diisopropylamino phosphine, bis benzyloxy phosphanyl diisopropylamine, phosphoramidous acid, bis 1-methylethyl-, bis phenylmethyl ester, n-dibenzyloxyphosphanyl-n-isopropyl-propan-2-amine, bis-bzl-2ipr-p, bis methylethyl bis phenylmethoxy phosphino amine
PubChem CID 196621
IUPAC Name N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OCC1=CC=CC=C1)OCC2=CC=CC=C2

Specifications

Density 1.03
Boiling Point 130°C (0.55 mmHg)
Flash Point 69°C (156°F)
Assay Percent Range 0.9
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 345.43
Percent Purity 95%
Chemical Name or Material Dibenzyl diisopropylphosphoramidite
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 629-628-9
TSCA No
storageNote1 Moisture Sensitive
Recommended Storage Keep cold

RUO – Research Use Only