missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl Dicarbonate 97.0+%, TCI America™
Supplier: TCI America P128125G
Specifications
| Dibenzyl Dicarbonate | |
| White | |
| MFCD00043124 | |
| Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester | |
| O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1 | |
| 286.28 | |
| 286.28 | |
| Crystalline Powder |
| 31139-36-3 | |
| C16H14O5 | |
| 25 g | |
| FHRRJZZGSJXPRQ-UHFFFAOYSA-N | |
| dibenzyl dicarbonate | |
| 3649378 | |
| ≥97.0% (T) |
Chemical Identifiers
| 31139-36-3 | |
| 286.28 | |
| FHRRJZZGSJXPRQ-UHFFFAOYSA-N | |
| 3649378 | |
| O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1 |
| C16H14O5 | |
| MFCD00043124 | |
| Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester | |
| dibenzyl dicarbonate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer