missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl Dicarbonate 97.0+%, TCI America™
$229.29 - $797.81
Chemical Identifiers
| CAS | 31139-36-3 |
|---|---|
| Molecular Formula | C16H14O5 |
| Molecular Weight (g/mol) | 286.28 |
| MDL Number | MFCD00043124 |
| InChI Key | FHRRJZZGSJXPRQ-UHFFFAOYSA-N |
| Synonym | Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester |
| PubChem CID | 3649378 |
| IUPAC Name | dibenzyl dicarbonate |
| SMILES | O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 31139-36-3 | |
| 286.28 | |
| FHRRJZZGSJXPRQ-UHFFFAOYSA-N | |
| 3649378 | |
| O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1 |
| C16H14O5 | |
| MFCD00043124 | |
| Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester | |
| dibenzyl dicarbonate |
Specifications
| 31139-36-3 | |
| C16H14O5 | |
| 5 g | |
| FHRRJZZGSJXPRQ-UHFFFAOYSA-N | |
| dibenzyl dicarbonate | |
| 3649378 | |
| ≥97.0% (T) | |
| Dibenzyl Dicarbonate |
| White | |
| MFCD00043124 | |
| Dibenzyl Pyrocarbonate, Pyrocarbonic Acid Dibenzyl Ester | |
| O=C(OCC1=CC=CC=C1)OC(=O)OCC1=CC=CC=C1 | |
| 286.28 | |
| 286.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer