missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl Carbonate 98.0+%, TCI America™
Supplier: TCI America C16005G
Specifications
| Dibenzyl Carbonate | |
| White-Yellow | |
| MFCD00014436 | |
| carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester | |
| O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1 | |
| 242.27 | |
| 242.27 | |
| Crystals |
| 3459-92-5 | |
| C15H14O3 | |
| 5 g | |
| PIZLBWGMERQCOC-UHFFFAOYSA-N | |
| dibenzyl carbonate | |
| 77002 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3459-92-5 | |
| 242.27 | |
| PIZLBWGMERQCOC-UHFFFAOYSA-N | |
| 77002 | |
| O=C(OCC1=CC=CC=C1)OCC1=CC=CC=C1 |
| C15H14O3 | |
| MFCD00014436 | |
| carbonic acid, bis phenylmethyl ester, benzyl carbonate, carbonic acid, dibenzyl ester, dibenzylcarbonate, carbonic acid dibenzyl ester, phenylmethyl phenylmethoxy formate, dibenzyl carbonate, acmc-209i8r, carbonic acid,bis phenylmethyl ester | |
| dibenzyl carbonate |
Safety and Handling
TSCA : Yes