missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl azodicarboxylate, 96%

Catalog No. AAL1934706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1934706 5 g
AAL1934714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1934706 Supplier Thermo Scientific Chemicals Supplier No. L1934706

CAS: 2449-05-0 | C16H14N2O4 | 298.298 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2449-05-0
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.298
MDL Number MFCD00016737
InChI Key IRJKSAIGIYODAN-ISLYRVAYSA-N
Synonym dibenzyl azodicarboxylate, azodicarboxylic acid dibenzyl ester, dbad, unii-o9nge39q8o, dibenzyl diazene-1,2-dicarboxylate, o9nge39q8o, diazenedicarboxylic acid, bis phenylmethyl ester, 1,2-diazenedicarboxylic acid, 1,2-bis phenylmethyl ester, phenylmethyl benzyloxycarbonyl diazenyl formate, dibenzylazodicarboxylate
PubChem CID 5387121
IUPAC Name benzyl (NE)-N-phenylmethoxycarbonyliminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)N=NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 43°C to 47°C
Flash Point >110°C (230°F)
Linear Formula C6H5CH2O2C-N=N-CO2CH2C6H5
Quantity 5 g
Beilstein 2298734
Sensitivity Light sensitive
Formula Weight 298.3
Percent Purity 96%
Chemical Name or Material Dibenzyl azodicarboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-508-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only