accessibility menu, dialog, popup

UserName

OONNOO(E/Z)

CAS RN 2449-05-0

IUPAC Name: N-{[(benzyloxy)carbonyl]imino}(benzyloxy)formamide
Synonyms: N-{[(benzyloxy)carbonyl]imino}benzyloxyformamide
Molecular Weight (g/mol): 298.3
Molecular Formula: C16H14N2O4
InChi Key: IRJKSAIGIYODAN-UHFFFAOYSA-N
SMILES: O=C(OCC1=CC=CC=C1)N=NC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)