missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl Adipate 95.0+%, TCI America™

Catalog No. A061825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A061825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A061825G Supplier TCI America Supplier No. A061825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2451-84-5
Molecular Formula C20H22O4
Molecular Weight (g/mol) 326.392
MDL Number MFCD00059667
InChI Key AEUORZZHALJMBM-UHFFFAOYSA-N
Synonym Adipic Acid Dibenzyl Ester
PubChem CID 75561
IUPAC Name dibenzyl hexanedioate
SMILES C1=CC=C(C=C1)COC(=O)CCCCC(=O)OCC2=CC=CC=C2

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 326.39
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material Dibenzyl Adipate