accessibility menu, dialog, popup

UserName

OOOO

CAS RN 2451-84-5

IUPAC Name: 1,6-dibenzyl hexanedioate
Synonyms: 1,6-dibenzyl hexanedioate
Molecular Weight (g/mol): 326.39
Molecular Formula: C20H22O4
InChi Key: AEUORZZHALJMBM-UHFFFAOYSA-N
SMILES: O=C(CCCCC(=O)OCC1=CC=CC=C1)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results