missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric Acid Monohydrate 98.0+%, TCI America™

Catalog No. p-7126692
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D135425G 25 g
D1354500G 500 g
2 options

Catalog No. D135425G

Supplier: TCI America D135425G

Only null left

Chemical Identifiers

CAS 62708-56-9
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149119
InChI Key YONLFQNRGZXBBF-OKILXGFUSA-N
Synonym 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
PubChem CID 44119738
IUPAC Name (2R,3S)-2,3-bis(benzoyloxy)butanedioic acid
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 90°C
Color White
Quantity 25 g
Formula Weight 358.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Dibenzoyl-L-tartaric Acid Monohydrate