missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid hydrate, 98%

Catalog No. p-7023105
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1618022 100 g
AAA1618036 500 g
AAA161800E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1618022 Supplier Thermo Scientific Chemicals Supplier No. A1618022

CAS: 62708-56-9 | C18H14O8 | 358.30 g/mol

(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.

Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 62708-56-9
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00149119
InChI Key YONLFQNRGZXBBF-OKILXGFUSA-N
Synonym 2r,3r-2,3-bis benzoyloxy succinic acid hydrate, --dibenzoyl-l-tartaric acid monohydrate, --dibenzoyl-l-tartaric acidmonohydrate, dibenzoyl-l---tartaric acid monohydrate, l--dibenzoyl-l-tartatic acid monohydrate, 2r,3r---dibenzoyl-l-tartaric acid monohydrate, 2r,3r-2,3-bis benzoyloxy butanedioic acid hydrate, dibenzoyl-l-tartaric acid monohydrate, l-dbta, pubchem7152
PubChem CID 44119738
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 90°C to 92°C
Solubility Information Soluble in methanol,acetone and organic solvents. Slightly soluble in water.
IUPAC Name (2R,3S)-2,3-dibenzoyloxybutanedioic acid;hydrate
Formula Weight 376.33 (358.31 Anhydrous)
Percent Purity ≥98%
Quantity 100 g
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid monohydrate
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 613-072-9
RTECSNumber WW8070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only