missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Dibenzoyl-L-tartaric acid hydrate, 98%
Description
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.
Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
(-)-Dibenzoyl-L-tartaric acid is used as an intermediate in pharmaceutical and agro chemical industries. It is used as chiral auxiliary in organic synthesis and as a resolution agent in the synthesis of rameleton.
Solubility
Soluble in methanol, acetone and organic solvents. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | 90°C to 92°C |
| Quantity | 2500 g |
| Solubility Information | Soluble in methanol,acetone and organic solvents. Slightly soluble in water. |
| IUPAC Name | (2R,3S)-2,3-bis(benzoyloxy)butanedioic acid |
| Formula Weight | 376.33 (358.31 Anhydrous) |
| Percent Purity | ≥98% |
| Chemical Name or Material | (-)-Dibenzoyl-L-tartaric acid monohydrate |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 613-072-9 |
| RTECSNumber | WW8070000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only