missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-Dibenzoyl-L-tartaric acid,98%
Description
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
It is sensitive to moisture. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 154°C to 156°C |
| Quantity | 100 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Slightly soluble in water. |
| IUPAC Name | (2R,3R)-2,3-bis(benzoyloxy)butanedioic acid |
| Formula Weight | 358.31 |
| Percent Purity | 99% |
| Chemical Name or Material | (-)-Dibenzoyl-L-tartaric acid, anhydrous |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 220-374-0 |
| RTECSNumber | WW8070000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only