missing translation for 'onlineSavingsMsg'
Learn More

(-)-Dibenzoyl-L-tartaric acid,98%

Catalog No. AAA1618114
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1618114 25 g
AAA1618122 100 g
AAA1618136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1618114 Supplier Thermo Scientific Chemicals Supplier No. A1618114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2743-38-6 | C18H14O8 | 358.30 g/mol

(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-Dibenzoyl-L-tartaric acid, anhydrous is used as a reagent used to produce chiral salts. Also used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
It is sensitive to moisture. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2743-38-6
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00003074
InChI Key YONLFQNRGZXBBF-ZIAGYGMSSA-N
Synonym dibenzoyl-l-tartaric acid, 2r,3r-2,3-bis benzoyloxy succinic acid, dibenzoyltartaric acid, --dibenzoyl-l-tartaric acid, dibenzoyl-l-tartaricacid, l-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2r,3r, unii-3q14b666vm, 2,3-bis benzoyloxy tartaric acid, o,o-dibenzoyl-+-tartaric acid
PubChem CID 75969
SMILES OC(=O)[C@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C to 156°C
Solubility Information Slightly soluble in water.
IUPAC Name (2R,3R)-2,3-dibenzoyloxybutanedioic acid
Formula Weight 358.31
Percent Purity 99%
Quantity 25 g
Sensitivity Hygroscopic
Chemical Name or Material (-)-Dibenzoyl-L-tartaric acid, anhydrous
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-374-0
RTECSNumber WW8070000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only