missing translation for 'onlineSavingsMsg'
Learn More

(+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%

Catalog No. p-7027753
Change view
Click to view available options
Quantity:
10 g
250 g
50 g
Catalog No. Quantity
AAB2475409 10 g
AAB2475418 50 g
AAB2475430 250 g
3 options

Catalog No. AAB2475409

Supplier: Thermo Scientific Chemicals B2475409

Only null left

CAS: 17026-42-5 | C18H14O8 | 358.30 g/mol

For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 17026-42-5
Molecular Formula C18H14O8
Molecular Weight (g/mol) 358.30
MDL Number MFCD00063222
InChI Key YONLFQNRGZXBBF-ZCWZLOQUNA-N
Synonym dibenzoyl-d-tartaric acid, +-dibenzoyl-d-tartaric acid, 2s,3s-2,3-bis benzoyloxy succinic acid, d-dibenzoyl tartaric acid, butanedioic acid, 2,3-bis benzoyloxy-, 2s,3s, +-o,o-dibenzoyltartaric acid, 2s,3s-+-dibenzoyltartaric acid, +-dibenzoyltartaric acid, +-dibenzoyl tartaric acid
PubChem CID 1550213
SMILES OC(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@H](OC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 154°C to 156°C
Beilstein 2227343
Solubility Information Soluble in ethanol. Insoluble in water.
IUPAC Name 2,3-bis(benzoyloxy)butanedioic acid
Formula Weight 358.31
Percent Purity 99%
Quantity 10 g
Optical Rotation +120° (c=5 in Methanol)
Chemical Name or Material (+)-Dibenzoyl-D-tartaric acid, anhydrous
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 241-097-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only