missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-Dibenzoyl-D-tartaric acid, anhydrous, 99%
Description
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
For chiral resolution(+)-Dibenzoyl-D-tartaric acid is used as a reagent in the preparation of chiral salts. It also serves as a reagent for chiral resolution of amino compounds.
Solubility
Soluble in ethanol. Insoluble in water.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 154°C to 156°C |
| Quantity | 10 g |
| Optical Rotation | +120° (c=5 in Methanol) |
| Beilstein | 2227343 |
| Solubility Information | Soluble in ethanol. Insoluble in water. |
| IUPAC Name | 2,3-bis(benzoyloxy)butanedioic acid |
| Formula Weight | 358.31 |
| Percent Purity | 99% |
| Chemical Name or Material | (+)-Dibenzoyl-D-tartaric acid, anhydrous |
Safety and Handling
| Hazard Category | H319 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P264b-P280i-P305+P351+P338 |
| EINECSNumber | 241-097-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only