missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberone 98.0+%, TCI America™

Catalog No. D180125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D180125G 25 g
D1801100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D180125G Supplier TCI America Supplier No. D180125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1210-35-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003587
InChI Key BMVWCPGVLSILMU-UHFFFAOYSA-N
Synonym dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
PubChem CID 14589
IUPAC Name tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-2-one
SMILES O=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Color White-Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 208.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dibenzosuberone
EINECSNumber (9)-2139
RTECSNumber HP1149700
TSCA Yes