missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberone, 97%

Catalog No. AAA1840514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1840514 25 g
AAA1840522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1840514 Supplier Thermo Scientific Chemicals Supplier No. A1840514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1210-35-1 | C15H12O | 208.26 g/mol

Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzosuberone is a potential antidepressant compound. It is used in the synthesis of amitriptyline.

Solubility
Soluble in water (0.03g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1210-35-1
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00003587
InChI Key BMVWCPGVLSILMU-UHFFFAOYSA-N
Synonym dibenzosuberone, dibenzocycloheptenone, dibenzosuberan-5-one, dibenzsuberone, 5-dibenzosuberone, 2,3:6,7-dibenzosuberone, 5h-dibenzo a,d cyclohepten-5-one, 10,11-dihydro, unii-8etk71th0h, dibenzo b,f cycloheptan-1-one, ccris 2780
PubChem CID 14589
SMILES O=C1C2=CC=CC=C2CCC2=CC=CC=C12

Specifications

Melting Point 32°C to 34°C
Density 1.156
Boiling Point 147°C to 149°C (0.3 mmHg)
Flash Point 200°C (392°F)
Refractive Index 1.634
Quantity 25 g
Beilstein 1454355
Solubility Information Soluble in water (0.03g/L at 20°C).
IUPAC Name 5,6-dihydrodibenzo[2,1-b
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material Dibenzosuberone
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 214-912-3
RTECSNumber HP1149700
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only