missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzofuran 98.0+%, TCI America™
Supplier: TCI America D0147100G
Specifications
| Dibenzofuran | |
| 83°C | |
| 288°C | |
| MFCD00004968 | |
| 3077 | |
| TXCDCPKCNAJMEE-UHFFFAOYSA-N | |
| dibenzofuran | |
| 568 | |
| 168.20 | |
| Crystalline Powder |
| 132-64-9 | |
| White | |
| C12H8O | |
| 100 g | |
| dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide | |
| C1=CC=C2C(=C1)C3=CC=CC=C3O2 | |
| 168.195 | |
| CHEBI:28145 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 132-64-9 | |
| 168.195 | |
| TXCDCPKCNAJMEE-UHFFFAOYSA-N | |
| 568 | |
| dibenzofuran |
| C12H8O | |
| MFCD00004968 | |
| dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide | |
| CHEBI:28145 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3O2 |
Safety and Handling
EINECSNumber : (9)-2295
RTECSNumber : HP4430000
TSCA : Yes