missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran 98.0+%, TCI America™

Catalog No. D014725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D014725G 25 g
D0147100G 100 g
D0147500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D014725G Supplier TCI America Supplier No. D014725G

Chemical Identifiers

CAS 132-64-9
Molecular Formula C12H8O
Molecular Weight (g/mol) 168.195
MDL Number MFCD00004968
InChI Key TXCDCPKCNAJMEE-UHFFFAOYSA-N
Synonym dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide
PubChem CID 568
ChEBI CHEBI:28145
IUPAC Name dibenzofuran
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3O2

Specifications

Melting Point 83°C
Color White
Boiling Point 288°C
Quantity 25 g
UN Number 3077
Formula Weight 168.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran
EINECSNumber (9)-2295
RTECSNumber HP4430000
TSCA Yes