missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran, 98%

Catalog No. AAA1652122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1652122 100 g
AAA1652114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1652122 Supplier Thermo Scientific Chemicals Supplier No. A1652122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 132-64-9 | C12H8O | 168.195 g/mol

Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Applications for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. Dibenzofuran as the model substrate in culture procedures.

Solubility
Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 132-64-9
Molecular Formula C12H8O
Molecular Weight (g/mol) 168.195
MDL Number MFCD00004968
InChI Key TXCDCPKCNAJMEE-UHFFFAOYSA-N
Synonym dibenzo b,d furan, diphenylene oxide, 2,2'-biphenylene oxide, 2,2'-biphenylylene oxide, dibenzofurans, dibenzol b,d furan, dibenzofurane, 1,1'-biphenyl-2,2'-diyl oxide
PubChem CID 568
ChEBI CHEBI:28145
IUPAC Name dibenzofuran
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3O2

Specifications

Melting Point 78°C to 83°C
Density 1.3
Boiling Point 287°C to 289°C
Flash Point 130°C (266°F)
Quantity 100 g
UN Number UN3077
Beilstein 121100
Solubility Information Insoluble in wate(<1mg/mL). Soluble in nonpolar organic solvents.
Formula Weight 168.2
Percent Purity 98%
Chemical Name or Material Dibenzofuran
Show More Show Less
Hazard Category H500
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 205-071-3
RTECSNumber HP4430000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only