missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzofuran-4-boronic Acid (contains varying amounts of Anhydride), TCI America™
$42.87 - $42.87
Chemical Identifiers
| CAS | 100124-06-9 |
|---|---|
| Molecular Formula | C12H9BO3 |
| MDL Number | MFCD00092336 |
| InChI Key | ZXHUJRZYLRVVNP-UHFFFAOYSA-N |
| PubChem CID | 2734328 |
Specifications
| 100124-06-9 | |
| White | |
| MFCD00092336 | |
| dibenzofuran-4-boronic acid, dibenzo b,d furan-4-ylboronic acid, 4-dibenzofuranyl boronic acid, 4-dibenzofuranboronic acid, 4-dibenzofuranboronicacid, 4-dibenzofuranylboronic acid, boronic acid, 4-dibenzofuranyl, dibenzo b,d furan-4-boronic acid, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaen-6-ylboronic acid | |
| B(C1=C2C(=CC=C1)C3=CC=CC=C3O2)(O)O | |
| 212.011 | |
| 212.01 | |
| Dibenzofuran-4-boronic Acid (contains varying amounts of Anhydride) |
| 258°C | |
| C12H9BO3 | |
| 1 g | |
| ZXHUJRZYLRVVNP-UHFFFAOYSA-N | |
| dibenzofuran-4-ylboronic acid | |
| 2734328 | |
| Crystalline Powder |
Safety and Handling
TSCA : No