missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. D47595G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47595G 5 g
D47591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47595G Supplier TCI America Supplier No. D47595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5397-82-0
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.21
MDL Number MFCD00136272
InChI Key OVJMIWIVPWPZMN-UHFFFAOYSA-N
Synonym dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
PubChem CID 220843
IUPAC Name 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
SMILES O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1

Specifications

Melting Point 74°C
Color White
Quantity 5 g
Formula Weight 196.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzofuran-2-carboxaldehyde