accessibility menu, dialog, popup

UserName

OO

CAS RN 5397-82-0

IUPAC Name: 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Synonyms: 8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-4-carbaldehyde
Molecular Weight (g/mol): 196.21
Molecular Formula: C13H8O2
InChi Key: OVJMIWIVPWPZMN-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)