missing translation for 'onlineSavingsMsg'
Learn More

Dibenzofuran-2-carboxaldehyde, 98%

Catalog No. p-7056143
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6490503 1 g
AAH6490506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6490503 Supplier Thermo Scientific Chemicals Supplier No. H6490503

CAS: 5397-82-0 | C13H8O2 | 196.21 g/mol

Used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an pharmaceutical intermediate.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 5397-82-0
Molecular Formula C13H8O2
Molecular Weight (g/mol) 196.21
MDL Number MFCD00136272
InChI Key OVJMIWIVPWPZMN-UHFFFAOYSA-N
Synonym dibenzo b,d furan-2-carbaldehyde, 2-dibenzofurancarboxaldehyde, dibenzofuran-2-carboxaldehyde, dibenzo b,d furan-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde, 2-dibenzofurancarbaldehyde, dibenzo b furan-2-carbaldehyde, benzo b benzofuran-2-carboxaldehyde, 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene-4-carbaldehyde, 8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-carbaldehyde
PubChem CID 220843
SMILES O=CC1=CC2=C(OC3=C2C=CC=C3)C=C1

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Beilstein 141044
Sensitivity Air Sensitive
IUPAC Name dibenzofuran-2-carbaldehyde
Formula Weight 196.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Dibenzofuran-2-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only