missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo[b,e][1,4]dioxine 99.0+%, TCI America™

Catalog No. D27761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27761G 1 g
D27765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27761G Supplier TCI America Supplier No. D27761G

Chemical Identifiers

CAS 262-12-4
Molecular Formula C12H8O2
Molecular Weight (g/mol) 184.19
MDL Number MFCD00022282
InChI Key NFBOHOGPQUYFRF-UHFFFAOYSA-N
Synonym oxanthrene, dibenzo b,e 1,4 dioxin, dibenzodioxin, phenodioxin, diphenylene dioxide, dibenzo 1,4 dioxin, dibenzo b,e 1,4 dioxine, dibenzo-para-dioxin
PubChem CID 9216
ChEBI CHEBI:28891
IUPAC Name oxanthrene
SMILES O1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 120°C
Color White
Quantity 1 g
Formula Weight 184.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo[b,e][1,4]dioxine
RTECSNumber HP3090000
TSCA Yes