missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo-24-crown 8-Ether 98.0+%, TCI America™
$152.86 - $434.32
Chemical Identifiers
| CAS | 14174-09-5 |
|---|---|
| Molecular Formula | C24H32O8 |
| Molecular Weight (g/mol) | 448.51 |
| MDL Number | MFCD00005101 |
| InChI Key | UNTITLLXXOKDTB-UHFFFAOYSA-N |
| PubChem CID | 84238 |
| IUPAC Name | 2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹â·]dotriaconta-1(32),12,14,16,28,30-hexaene |
| SMILES | C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1 |
Chemical Identifiers
| 14174-09-5 | |
| 448.51 | |
| UNTITLLXXOKDTB-UHFFFAOYSA-N | |
| 2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹â·]dotriaconta-1(32),12,14,16,28,30-hexaene |
| C24H32O8 | |
| MFCD00005101 | |
| 84238 | |
| C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1 |
Specifications
| 14174-09-5 | |
| White-Yellow | |
| MFCD00005101 | |
| UNTITLLXXOKDTB-UHFFFAOYSA-N | |
| 2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹â·]dotriaconta-1(32),12,14,16,28,30-hexaene | |
| 84238 | |
| ≥98.0% (GC) | |
| Dibenzo-24-crown 8-Ether |
| 104°C | |
| C24H32O8 | |
| 1 g | |
| C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1 | |
| 448.51 | |
| 448.51 | |
| Crystalline Powder |
Safety and Handling
TSCA : No