CAS RN 14174-09-5
CAS RN 14174-09-5
IUPAC Name:
2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹⁷]dotriaconta-1(32),12,14,16,28,30-hexaene
Synonyms:
2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹⁷]dotriaconta-1(32),12,14,16,28,30-hexaene
Molecular Weight (g/mol):
448.51
Molecular Formula:
C24H32O8
InChi Key:
UNTITLLXXOKDTB-UHFFFAOYSA-N
SMILES:
C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1
1
–
1
of
1
results
1
Dibenzo-24-crown 8-Ether 98.0+%, TCI America™
CAS: 14174-09-5 Molecular Formula: C24H32O8 Molecular Weight (g/mol): 448.51 MDL Number: MFCD00005101 InChI Key: UNTITLLXXOKDTB-UHFFFAOYSA-N PubChem CID: 84238 IUPAC Name: 2,5,8,11,18,21,24,27-octaoxatricyclo[26.4.0.0¹²,¹⁷]dotriaconta-1(32),12,14,16,28,30-hexaene SMILES: C1COCCOC2=CC=CC=C2OCCOCCOCCOC2=CC=CC=C2OCCO1