missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzo-18-crown 6-Ether 99.0+%, TCI America™
Supplier: TCI America D153325G
Specifications
| Dibenzo-18-crown 6-Ether | |
| 163°C | |
| C20H24O6 | |
| 25 g | |
| YSSSPARMOAYJTE-UHFFFAOYSA-N | |
| 360.406 | |
| CHEBI:358732 | |
| ≥99.0% (GC) |
| 14187-32-7 | |
| 382°C | |
| MFCD00005098 | |
| dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene | |
| C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1 | |
| 26541 | |
| 360.41 | |
| Crystalline Powder |
Chemical Identifiers
| 14187-32-7 | |
| 360.406 | |
| YSSSPARMOAYJTE-UHFFFAOYSA-N | |
| 26541 | |
| C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1 |
| C20H24O6 | |
| MFCD00005098 | |
| dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene | |
| CHEBI:358732 |
Safety and Handling
RTECSNumber : HP5386000
TSCA : Yes