missing translation for 'onlineSavingsMsg'
Learn More

Dibenzo-18-crown 6-Ether 99.0+%, TCI America™

Catalog No. p-7126773
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15335G 5 g
D153325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15335G Supplier TCI America Supplier No. D15335G
Only null left

Chemical Identifiers

CAS 14187-32-7
Molecular Formula C20H24O6
Molecular Weight (g/mol) 360.406
MDL Number MFCD00005098
InChI Key YSSSPARMOAYJTE-UHFFFAOYSA-N
Synonym dibenzo-18-crown-6, dibenzo-18-crown-6-ether, crown 18, 6,7,9,10,17,18,20,21-octahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dibenzo-18-crown, unii-0a7w45jcs9, 2,3:11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadecane, dibenzo-18-crown-6 ether, dibenzo-18-crown 6-ether, 2,3,11,12-dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene
PubChem CID 26541
ChEBI CHEBI:358732
SMILES C1COC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

Specifications

Melting Point 163°C
Boiling Point 382°C
Quantity 5 g
Formula Weight 360.41
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dibenzo-18-crown 6-Ether
RTECSNumber HP5386000
TSCA Yes